Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
twk-36
Entrez Gene ID
180164
Gene Name
TWiK family of potassium channels
Gene Aliases
CELE_R12G8.2
Chromosome Location
Chr.V: 17743651 - 17747059 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075084.4 NP_507485.2 4 - 5 863 108

View Full Product Details