Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pgp-9
Entrez Gene ID
180165
Gene Name
P-GlycoProtein related
Gene Aliases
CELE_C47A10.1
Chromosome Location
Chr.V: 17755887 - 17764751 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075086.5 NP_507487.1 1 - 2 53 77

View Full Product Details