Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
csn-1
Entrez Gene ID
180168
Gene Name
COP9 signalosome complex subunit 1
Gene Aliases
CELE_Y59A8A.1
Chromosome Location
Chr.V: 17861368 - 17871193 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383483.1 2 - 3 866 114

View Full Product Details