Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
par-4
Entrez Gene ID
180185
Gene Name
protein-coding
Gene Aliases
CELE_Y59A8B.14
Chromosome Location
Chr.V: 18098676 - 18106868 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380857.1 NP_001366977.1 6 - 7 1030 65
NM_001380858.1 NP_001367285.1 6 - 7 1007 65
NM_001380859.1 NP_001367284.1 3 - 4 596 65
GenBank mRNA AF160189.1 - 6 - 7 1053 65

View Full Product Details