Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srg-34
Entrez Gene ID
190136
Gene Name
Serpentine receptor class gamma
Gene Aliases
CELE_Y51A2D.12
Chromosome Location
Chr.V: 18562179 - 18564886 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075230.3 NP_507631.1 2 - 3 400 121

View Full Product Details