Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sdz-6
Entrez Gene ID
183410
Gene Name
SKN-1 Dependent Zygotic transcript
Gene Aliases
CELE_C43D7.5
Chromosome Location
Chr.V: 19320613 - 19321351 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075363.2 NP_507764.1 1 - 2 314 126

View Full Product Details