Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
srw-84
Entrez Gene ID
189867
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_Y43F8A.4
Chromosome Location
Chr.V: 19381268 - 19384760 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075371.1 NP_507772.1 2 - 3 579 149

View Full Product Details