Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gly-4
Entrez Gene ID
180302
Gene Name
protein-coding
Gene Aliases
CELE_Y116F11B.12
Chromosome Location
Chr.V: 19881372 - 19902647 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029045.5 NP_001024216.1 11 - 12 2194 86
GenBank mRNA AF031834.1 - 8 - 9 1746 86

View Full Product Details