Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sec-23
Entrez Gene ID
180317
Gene Name
Protein transport protein SEC23
Gene Aliases
CELE_Y113G7A.3
Chromosome Location
Chr.V: 20091263 - 20105693 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075476.7 NP_507877.1 5 - 6 589 67

View Full Product Details