Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srbc-34
Entrez Gene ID
190974
Gene Name
Serpentine Receptor, class BC (class B-like)
Gene Aliases
CELE_Y113G7B.9
Chromosome Location
Chr.V: 20206318 - 20207907 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075499.3 NP_507900.2 2 - 3 523 157

View Full Product Details