Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
grd-11
Entrez Gene ID
186880
Gene Name
GRounDhog (hedgehog-like family)
Gene Aliases
CELE_K02E2.2
Chromosome Location
Chr.V: 20356772 - 20365773 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075522.4 NP_507923.2 14 - 15 3011 91

View Full Product Details