Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cnc-7
Entrez Gene ID
180337
Gene Name
CaeNaCin (Caenorhabditis bacteriocin)
Gene Aliases
CELE_F53H2.2
Chromosome Location
Chr.V: 20394073 - 20394735 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075529.5 NP_507930.1 1 - 2 74 63

View Full Product Details