Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ife-2
Entrez Gene ID
180393
Gene Name
Eukaryotic translation initiation factor 4E-2
Gene Aliases
CELE_R04A9.4
Chromosome Location
Chr.X: 381338 - 382709 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075693.5 NP_508094.1 1 - 2 79 69

View Full Product Details