Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
che-2
Entrez Gene ID
180401
Gene Name
Uncharacterized protein
Gene Aliases
CELE_F38G1.1
Chromosome Location
Chr.X: 481450 - 486504 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075705.6 NP_508106.1 11 - 12 1838 77
GenBank mRNA AJ011523.1 - 11 - 12 1860 77

View Full Product Details