Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lgc-32
Entrez Gene ID
188603
Gene Name
Neur_chan_LBD domain-containing protein
Gene Aliases
CELE_T19D7.1
Chromosome Location
Chr.X: 673021 - 676484 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075738.4 NP_508139.2 5 - 6 719 70

View Full Product Details