Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nas-15
Entrez Gene ID
180426
Gene Name
Zinc metalloproteinase nas-15
Gene Aliases
CELE_T04G9.2
Chromosome Location
Chr.X: 777721 - 780533 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075753.8 NP_508154.2 1 - 2 120 89

View Full Product Details