Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
daf-3
Entrez Gene ID
180431
Gene Name
protein-coding
Gene Aliases
CELE_F25E2.5
Chromosome Location
Chr.X: 817922 - 825475 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029433.7 NP_001024604.1 4 - 5 574 87
NM_001029434.5 NP_001024605.1 2 - 3 346 87
NM_001313412.4 NP_001300341.1 5 - 5 629 87
NM_001313413.4 NP_001300342.1 4 - 5 576 87
NM_001313414.3 NP_001300343.1 4 - 5 576 87
NM_001313415.4 NP_001300344.1 2 - 3 293 87
NM_001380898.1 NP_001368578.1 2 - 3 293 87
NM_075760.6 NP_508161.3 5 - 6 678 87
GenBank mRNA AF005205.1 - 2 - 3 341 87

View Full Product Details