Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sel-12
Entrez Gene ID
180441
Gene Name
Presenilin sel-12
Gene Aliases
CELE_F35H12.3
Chromosome Location
Chr.X: 915867 - 918290 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075774.7 NP_508175.1 4 - 5 779 80

View Full Product Details