Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rme-6
Entrez Gene ID
180463
Gene Name
protein-coding
Gene Aliases
CELE_F49E7.1
Chromosome Location
Chr.X: 1097146 - 1103324 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383525.1 14 - 15 2721 62
NM_075807.5 NP_508208.2 14 - 15 2763 62
GenBank mRNA AY750054.1 - 14 - 15 2763 62

View Full Product Details