Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dop-4
Entrez Gene ID
183715
Gene Name
Dopamine receptor 4
Gene Aliases
CELE_C52B11.3
Chromosome Location
Chr.X: 1295253 - 1298830 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075837.5 NP_508238.2 6 - 7 1114 66

View Full Product Details