Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dhs-25
Entrez Gene ID
180486
Gene Name
DeHydrogenases, Short chain
Gene Aliases
CELE_F09E10.3
Chromosome Location
Chr.X: 1486473 - 1489326 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075881.6 NP_508282.2 2 - 3 380 80

View Full Product Details