Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxa-52
Entrez Gene ID
184153
Gene Name
FTH domain-containing protein
Gene Aliases
CELE_F07G6.6
Chromosome Location
Chr.X: 1710149 - 1712638 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075913.4 NP_508314.2 2 - 3 716 134

View Full Product Details