Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rgs-9
Entrez Gene ID
180506
Gene Name
Regulator of G-protein signaling rgs-9
Gene Aliases
CELE_ZC53.7
Chromosome Location
Chr.X: 1915258 - 1918552 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075949.7 NP_508350.1 1 - 2 253 108

View Full Product Details