Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
F52D2.6
Entrez Gene ID
186105
Gene Name
Uncharacterized protein
Gene Aliases
CELE_F52D2.6
Chromosome Location
Chr.X: 1975635 - 1984841 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075964.6 NP_508365.2 4 - 5 986 91

View Full Product Details