Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hex-1
Entrez Gene ID
180533
Gene Name
Beta-hexosaminidase A
Gene Aliases
CELE_T14F9.3
Chromosome Location
Chr.X: 2221150 - 2226620 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076008.7 NP_508409.1 7 - 8 1028 74
GenBank mRNA AM748820.1 - 6 - 7 913 74

View Full Product Details