Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hex-1
Entrez Gene ID
180533
Gene Name
Beta-hexosaminidase A
Gene Aliases
CELE_T14F9.3
Chromosome Location
Chr.X: 2221150 - 2226620 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076008.7 NP_508409.1 10 - 11 1591 110
GenBank mRNA AM748820.1 - 9 - 10 1476 110

View Full Product Details