Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
syg-1
Entrez Gene ID
180555
Gene Name
protein-coding
Gene Aliases
CELE_K02E10.8
Chromosome Location
Chr.X: 2509313 - 2515324 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129687.6 NP_001123159.1 9 - 10 1228 73
NM_001373246.4 NP_001360786.1 10 - 10 1229 73
NM_076056.8 NP_508457.3 9 - 10 1222 73

View Full Product Details