Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
igcm-3
Entrez Gene ID
180569
Gene Name
protein-coding
Gene Aliases
CELE_T02C5.3
Chromosome Location
Chr.X: 2698257 - 2705859 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129703.4 NP_001123175.1 9 - 10 1601 91
NM_076078.6 NP_508479.2 9 - 10 1601 91

View Full Product Details