Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
twk-16
Entrez Gene ID
180594
Gene Name
TWiK family of potassium channels
Gene Aliases
CELE_F52E4.4
Chromosome Location
Chr.X: 3081125 - 3085401 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076125.4 NP_508526.2 1 - 2 67 89
GenBank mRNA AF083645.1 - 1 - 2 59 89

View Full Product Details