Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
wrt-2
Entrez Gene ID
180597
Gene Name
WaRThog (hedgehog-like family)
Gene Aliases
CELE_F52E4.6
Chromosome Location
Chr.X: 3092395 - 3094448 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076128.6 NP_508529.1 7 - 8 1122 151

View Full Product Details