The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029687.6 | NP_001024858.2 | 6 - 7 | 776 | 88 |
NM_076226.6 | NP_508627.5 | 5 - 6 | 678 | 88 |