Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tag-52
Entrez Gene ID
180657
Gene Name
Putative protein tag-52
Gene Aliases
CELE_C02F12.4
Chromosome Location
Chr.X: 3674041 - 3678557 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076232.4 NP_508633.1 5 - 6 978 90

View Full Product Details