Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tag-18
Entrez Gene ID
180662
Gene Name
Uncharacterized protein
Gene Aliases
CELE_T14G12.3
Chromosome Location
Chr.X: 3731894 - 3745470 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076241.9 NP_508642.1 4 - 5 490 123

View Full Product Details