Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lim-4
Entrez Gene ID
180672
Gene Name
protein-coding
Gene Aliases
CELE_ZC64.4
Chromosome Location
Chr.X: 3869404 - 3873545 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001351878.2 NP_001338857.1 - 1077 71
NM_001351879.2 NP_001338856.1 - 1233 71
NM_076268.2 NP_508669.1 7 - 8 1259 71
GenBank mRNA U72348.1 - 7 - 8 1010 71

View Full Product Details