Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srv-5
Entrez Gene ID
185979
Gene Name
protein-coding
Gene Aliases
CELE_F48D6.2
Chromosome Location
Chr.X: 4252347 - 4254174 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029517.3 NP_001024688.1 2 - 3 299 79
NM_001029518.3 NP_001024689.1 2 - 3 290 79

View Full Product Details