Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dop-1
Entrez Gene ID
180714
Gene Name
protein-coding
Gene Aliases
CELE_F15A8.5
Chromosome Location
Chr.X: 4411301 - 4418851 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029407.4 NP_001024578.1 4 - 5 551 80
NM_001029408.5 NP_001024579.1 3 - 4 462 80
NM_001383561.1 4 - 5 1161 80

View Full Product Details