Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
asb-2
Entrez Gene ID
180718
Gene Name
ATP Synthase B homolog
Gene Aliases
CELE_F02E8.1
Chromosome Location
Chr.X: 4469766 - 4471480 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076369.5 NP_508770.1 2 - 3 281 64

View Full Product Details