Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ceh-1
Entrez Gene ID
191614
Gene Name
Homeobox protein ceh-1
Gene Aliases
CELE_F16H11.4
Chromosome Location
Chr.X: 4653163 - 4654157 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076395.6 NP_508796.2 3 - 4 406 110

View Full Product Details