Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hyl-2
Entrez Gene ID
180743
Gene Name
Ceramide synthase hyl-2
Gene Aliases
CELE_K02G10.6
Chromosome Location
Chr.X: 4696754 - 4699461 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076402.9 NP_508803.3 6 - 7 874 88

View Full Product Details