Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
apl-1
Entrez Gene ID
180783
Gene Name
protein-coding
Gene Aliases
CELE_C42D8.8
Chromosome Location
Chr.X: 5112416 - 5116868 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076469.6 NP_508870.3 3 - 4 368 64
NM_076470.7 NP_508871.1 3 - 4 370 64

View Full Product Details