Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- bus-8
- Entrez Gene ID
-
180821
- Gene Name
- Uncharacterized protein
- Gene Aliases
-
CELE_T23F2.1
- Chromosome Location
-
Chr.X: 5492915 - 5496053 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_076531.6 |
NP_508932.1 |
3 - 4 |
582 |
96 |
GenBank mRNA |
EU732660.1 |
- |
3 - 4 |
521 |
96 |
- Gene Symbol
- T23F2.13
- Entrez Gene ID
-
36805045
- Gene Name
- Uncharacterized protein
- Gene Aliases
-
CELE_T23F2.13
- Chromosome Location
-
Chr.X: 5492915 - 5496013 on Build WBcel235
View Full Product Details