Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
egl-13
Entrez Gene ID
180833
Gene Name
protein-coding
Gene Aliases
CELE_T22B7.1
Chromosome Location
Chr.X: 5663835 - 5674067 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029747.6 NP_001024918.1 8 - 9 951 84
NM_001307913.5 NP_001294842.1 5 - 6 1202 84
NM_001330899.4 NP_001317909.1 6 - 7 778 84
NM_001330900.4 NP_001317910.1 2 - 3 403 84
NM_001380981.1 NP_001368074.1 6 - 7 730 84
NM_001383586.1 7 - 8 832 84
NR_138511.1 7 - 8 1003 84
NR_138512.1 7 - 8 1167 84
GenBank mRNA AF097319.1 - 7 - 8 882 84

View Full Product Details