Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hsp-25
Entrez Gene ID
180872
Gene Name
protein-coding
Gene Aliases
CELE_C09B8.6
Chromosome Location
Chr.X: 6034463 - 6041691 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029203.3 NP_001024374.1 4 - 5 600 83
NM_001380987.1 NP_001367469.1 4 - 5 483 83
NM_001383593.1 4 - 5 2471 83

View Full Product Details