Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ggr-2
Entrez Gene ID
191637
Gene Name
protein-coding
Gene Aliases
CELE_C45B2.4
Chromosome Location
Chr.X: 6070497 - 6081881 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029301.6 NP_001024472.2 6 - 7 796 89
NM_001380988.1 NP_001368738.1 5 - 6 658 89
NM_001380989.1 NP_001368736.1 5 - 6 658 89
NM_001380990.1 NP_001368737.1 5 - 6 658 89
NM_001380991.1 NP_001368739.1 5 - 6 658 89
NM_001380992.2 NP_001368740.1 5 - 6 728 89
NM_001380993.1 NP_001368743.1 5 - 6 658 89
NM_001380994.1 NP_001368744.1 5 - 6 658 89
NM_001380995.1 NP_001368741.1 5 - 6 658 89
NM_001380996.1 NP_001368742.1 5 - 6 658 89
NM_001380997.1 NP_001368745.1 5 - 6 658 89
NM_001380998.2 NP_001368746.1 5 - 6 731 89
NM_001380999.1 NP_001367470.1 5 - 6 658 89

View Full Product Details