Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nnt-1
Entrez Gene ID
180884
Gene Name
Nicotinamide Nucleotide Transhydrogenase
Gene Aliases
CELE_C15H9.1
Chromosome Location
Chr.X: 6122311 - 6127554 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076627.6 NP_509028.1 15 - 16 2630 90

View Full Product Details