Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mom-1
Entrez Gene ID
180904
Gene Name
Protein-serine O-palmitoleoyltransferase porcupine
Gene Aliases
CELE_T07H6.2
Chromosome Location
Chr.X: 6293124 - 6298305 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076653.5 NP_509054.1 3 - 4 598 108
GenBank mRNA AF013489.1 - 3 - 4 595 108

View Full Product Details