Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cdf-1
Entrez Gene ID
180923
Gene Name
protein-coding
Gene Aliases
CELE_C15B12.7
Chromosome Location
Chr.X: 6499916 - 6505133 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076695.6 NP_509096.1 2 - 3 388 130

View Full Product Details