Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glr-8
Entrez Gene ID
180924
Gene Name
protein-coding
Gene Aliases
CELE_F22A3.3
Chromosome Location
Chr.X: 6505980 - 6509957 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001381009.1 NP_001368711.1 3 - 4 341 119
NM_001381010.2 NP_001367505.1 3 - 4 637 119

View Full Product Details