Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ilys-5
Entrez Gene ID
180928
Gene Name
protein-coding
Gene Aliases
CELE_F22A3.6
Chromosome Location
Chr.X: 6533493 - 6536840 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029423.7 NP_001024594.1 3 - 4 324 64
NM_001313410.4 NP_001300339.1 2 - 2 186 64
NR_132543.1 2 - 3 1003 64

View Full Product Details