Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
acr-3
Entrez Gene ID
180937
Gene Name
Acetylcholine receptor subunit beta-type acr-3
Gene Aliases
CELE_K11G12.7
Chromosome Location
Chr.X: 6709538 - 6712925 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076728.4 NP_509129.1 13 - 14 1396 109
GenBank mRNA Y08637.1 - - 1396 109

View Full Product Details